About This Item
assay
99.9% trace metals basis
form
chips
reaction suitability
reagent type: catalyst
core: thulium
resistivity
90 μΩ-cm, 20°C
bp
1950 °C (lit.)
mp
1545 °C (lit.)
density
9.332 g/mL at 25 °C (lit.)
SMILES string
[Tm]
InChI
1S/Tm
InChI key
FRNOGLGSGLTDKL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Sol. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
4.1B - Flammable solid hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service