Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

260185

Sigma-Aldrich

3,3-Dimethyl-1,2-butanediol

technical grade, ≥85%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH(OH)CH2OH
CAS Number:
Molecular Weight:
118.17
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

assay

≥85%

form

solid

mp

37-39 °C (lit.)

SMILES string

CC(C)(C)C(O)CO

InChI

1S/C6H14O2/c1-6(2,3)5(8)4-7/h5,7-8H,4H2,1-3H3

InChI key

YVHAOWGRHCPODY-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

210.2 °F - closed cup

flash_point_c

99.00 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service