Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

25886

Sigma-Aldrich

3-(4-Quinolyl)-DL-alanine dihydrochloride dihydrate

≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O2 · 2HCl · 2H2O
Molecular Weight:
325.19
Beilstein/REAXYS Number:
3736980
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98.0% (HPLC)

application(s)

peptide synthesis

SMILES string

O.O.Cl.Cl.NC(Cc1ccnc2ccccc12)C(O)=O

InChI

1S/C12H12N2O2.2ClH.2H2O/c13-10(12(15)16)7-8-5-6-14-11-4-2-1-3-9(8)11;;;;/h1-6,10H,7,13H2,(H,15,16);2*1H;2*1H2

InChI key

RXCRMUQUBPESMX-UHFFFAOYSA-N

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service