Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

257915

Sigma-Aldrich

Trimethyltin bromide

95%

Synonym(s):

Bromotrimethyltin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SnBr
CAS Number:
Molecular Weight:
243.72
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

mp

26-27 °C (lit.)

SMILES string

C[Sn](C)(C)Br

InChI

1S/3CH3.BrH.Sn/h3*1H3;1H;/q;;;;+1/p-1

InChI key

MZGUIAFRJWSYJJ-UHFFFAOYSA-M

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Aldrichimica Acta, 17, 75-75 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service