Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

257893

Sigma-Aldrich

Tributyltin bromide

technical grade, 90%

Synonym(s):

Bromotributyltin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3SnBr
CAS Number:
Molecular Weight:
369.96
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

technical grade

assay

90%

refractive index

n20/D 1.507 (lit.)

bp

120-122 °C/2 mmHg (lit.)

density

1.338 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](Br)(CCCC)CCCC

InChI

1S/3C4H9.BrH.Sn/c3*1-3-4-2;;/h3*1,3-4H2,2H3;1H;/q;;;;+1/p-1

InChI key

FVRKTAOFDKFAMI-UHFFFAOYSA-M

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Girish K Srivastava et al.
Regulatory toxicology and pharmacology : RTP, 110, 104527-104527 (2019-11-17)
Perfluorocarbon liquids (PFCLs) have been considered safe for intraocular manipulation of the retina, but since 2013 many cases of acute eye toxicity cousing blindness have been reported in various countries when using various commercial PFCLs. All these PFCLs were CE
Luba Hunakova et al.
Toxicology letters, 279, 16-21 (2017-07-16)
Among a variety of metal containing organic compounds, tin derivatives are enjoying an increasing interest and appear to be very promising as potential drug candidates. We studied eight organometallic derivatives, nuclear retinoid X receptor (RXR) ligands and two germanium containing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service