Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

257656

Sigma-Aldrich

Ethyl 3-thiopheneacetate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10O2S
CAS Number:
Molecular Weight:
170.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

liquid

refractive index

n20/D 1.509 (lit.)

bp

97-98 °C/8 mmHg (lit.)

density

1.137 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)Cc1ccsc1

InChI

1S/C8H10O2S/c1-2-10-8(9)5-7-3-4-11-6-7/h3-4,6H,2,5H2,1H3

InChI key

FZBNQIWPYCUPAP-UHFFFAOYSA-N

General description

Ethyl 3-thiopheneacetate undergoes copolymerization with 3-alkylthiophene to yield poly(3-alkyl-2,5-thienylene-co-3-methyiene-ethylcarboxylate-2,5-thienylene)[1].

Application

Ethyl 3-thiopheneacetate has been used in preparation of:
  • solution-processible hybrid polymers, consisting of a polythiophene main chain and randomly distributed alkyl and oligoaniline side groups[2]
  • poly(thiophene-3-acetic acid)[3]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

217.4 °F - closed cup

flash_point_c

103.00 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Random and regioregular thiophene-based copolymers containing oligoaniline side chains: Synthesis, spectroscopic and spectroelectrochemical investigations.
Buga K, et al.
Synthetic Metals, 153(1-3), 137-140 (2005)
Postpolymerization grafting of aniline tetramer on polythiophene chain: Structural organization of the product and its electrochemical and spectroelectrochemical properties.
Buga K, et al.
Chemistry of Materials, 17(23), 5754-5762 (2005)
Construction of organic-inorganic hybrid ultrathin films self-assembled from poly (thiophene-3-acetic acid) and TiO2.
Ding H, et al.
Journal of Materials Chemistry, 12(12), 3585-3590 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service