Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

257648

Sigma-Aldrich

Ethyl 2-thiopheneacetate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10O2S
CAS Number:
Molecular Weight:
170.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

liquid

refractive index

n20/D 1.51 (lit.)

bp

119-121 °C/23 mmHg (lit.)

density

1.134 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)Cc1cccs1

InChI

1S/C8H10O2S/c1-2-10-8(9)6-7-4-3-5-11-7/h3-5H,2,6H2,1H3

InChI key

QSUANHXENVRFDN-UHFFFAOYSA-N

Application

Ethyl 2-thiopheneacetate has been used in preparation of:
  • S-(2-(5-((amidinothio)methyl)-2-thienyl)ethyl)isothiourea[1]
  • pyrrolo [3,2-b] pyrrole-based copolymers for organic photovoltaic device[2]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

219.2 °F - closed cup

flash_point_c

104.00 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrrolo [3, 2-b] pyrrole-Based Copolymers as Donor Materials for Organic Photovoltaics.
Song S, et al.
Bull. Korean Chem. Soc., 34(11), 3399-3399 (2013)
E P Garvey et al.
The Journal of biological chemistry, 269(43), 26669-26676 (1994-10-28)
S-Ethylisothiourea was a potent competitive inhibitor of human nitric oxide synthase (NOS), with Ki values of 17, 36, and 29 nM for the inducible (i), endothelial (e), and neuronal (n) isozymes, respectively. Unlike some potent inhibitors of NOS, no time

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service