Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

257249

Sigma-Aldrich

Tripropylaluminum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)3Al
CAS Number:
Molecular Weight:
156.24
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

bp

82-84 °C/2 mmHg (lit.)

mp

−107 °C (lit.)

density

0.823 g/mL at 25 °C (lit.)

SMILES string

CCC[Al](CCC)CCC

InChI

1S/3C3H7.Al/c3*1-3-2;/h3*1,3H2,2H3;

InChI key

CNWZYDSEVLFSMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Studies of the kinetics and mechanism of Zr-catalyzed chain growth
  • Copper-catalyzed conjugate addition to nitrodienes and nitroenynes or oxabicyclohelptadienedicarboxylates
  • Copper-catalyzed stereoselective ring opening / desymmetrization of polycyclic meso-hydrazines or 1,4-dihydro-1,4-naphthalene epoxide
  • Synthesis of β-hydroxylactams via alkylative aldol cyclization with α,β-unsaturated carboxamides

Recommended products

Angled septum-inlet adapter Z118206 or straight septum-inlet adapter, 6mm I.D. inlet, Z118141 or straight septum-inlet adapter, 13mm I.D. inlet, Z118192 is recommended.

septum inlet adapter

Product No.
Description
Pricing

pictograms

FlameCorrosion

signalword

Danger

Hazard Classifications

Pyr. Liq. 1 - Skin Corr. 1B - Water-react 1

supp_hazards

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service