Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

256870

Sigma-Aldrich

Diisobutylaluminum hydride solution

1.0 M in toluene

Synonym(s):

DIBAL, DIBAL-H

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHCH2]2AlH
CAS Number:
Molecular Weight:
142.22
Beilstein/REAXYS Number:
4123663
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

reaction suitability

reagent type: reductant

concentration

1.0 M in toluene

density

0.858 g/mL at 25 °C

SMILES string

CC(C)C[AlH]CC(C)C

InChI

1S/2C4H9.Al.H/c2*1-4(2)3;;/h2*4H,1H2,2-3H3;;

InChI key

AZWXAPCAJCYGIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in Pd-catalyzed reductive debromination of secondary alkyl bromides. O-Debenzylation and ring opening of perbenzylated furanosides. Convenient in situ generation of HZrCp2Cl from ZrCp2Cl2 and DIBAL-H.

Recommended products

Angled septum-inlet adapter Z118206 or straight septum-inlet adapter, 6mm I.D. inlet, Z118141 or straight septum-inlet adapter, 13mm I.D. inlet, Z118192 is recommended.

septum inlet adapter

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D J Kopecky et al.
The Journal of organic chemistry, 65(1), 191-198 (2000-05-18)
An optimized protocol for the DIBALH reductive acetylation of acyclic esters and diesters is described. This reductive acetylation procedure allows a wide variety of esters to be converted into the corresponding alpha-acetoxy ethers in good to excellent yields. It was
J Marco-Contelles et al.
Carbohydrate research, 335(1), 63-70 (2001-09-13)
The reaction of DIBALH with bis(heteroannulated)-pyranosides containing the perhydrofuro[2,3-b]pyran moiety is described. The hydride attack at the anomeric carbon (C-9a) resulted in the exclusive tetrahydrofuran ring opening. The selectivity of this reaction has been evaluated as other benzylidene acetals built
Hidetsura Cho et al.
The Journal of organic chemistry, 75(3), 627-636 (2009-12-31)
A systematic investigation of the reductive ring-expansion reaction of cyclic ketoximes fused to aromatic rings with diisobutylaluminum hydride (DIBALH) is described. This reaction regioselectively afforded a variety of five- to eight-membered bicyclic heterocycles or tricyclic heterocycles containing nitrogen neighboring an
Y Kitade et al.
Nucleic acids symposium series, (27)(27), 107-108 (1992-01-01)
Reaction of purine nucleosides, such as 2',3'-isopropylideneinosine (1a) and 2',3'-isopropylideneadenosine (1c), with diisobutylaluminum hydride (DIBAL) in dry tetrahydrofurane resulted in the formation of the corresponding 9-(2',3'-isopropylideneribity)purines (2) in good yields. Oxidation of the ribityl derivatives (2) with NalO4 and subsequent
Damien Webb et al.
Organic letters, 14(2), 568-571 (2011-12-31)
A continuous flow system for the multiparameter (flow rate, temperature, residence time, stoichiometry) optimization of the DIBALH reduction of esters to aldehydes is described. Incorporating an in-line quench (MeOH), these transformations are generally complete in fewer than 60 s. Mixing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service