Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

256854

Sigma-Aldrich

Diisobutylaluminum hydride solution

1.0 M in hexanes

Synonym(s):

DIBAL, DIBAL-H

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHCH2]2AlH
CAS Number:
Molecular Weight:
142.22
Beilstein/REAXYS Number:
4123663
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

form

liquid

reaction suitability

reagent type: reductant

concentration

1.0 M in hexanes

density

0.701 g/mL at 25 °C

SMILES string

CC(C)C[AlH]CC(C)C

InChI

1S/2C4H9.Al.H/c2*1-4(2)3;;/h2*4H,1H2,2-3H3;;

InChI key

AZWXAPCAJCYGIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in Pd-catalyzed reductive debromination of secondary alkyl bromides. O-Debenzylation and ring opening of perbenzylated furanosides. Convenient in situ generation of HZrCp2Cl from ZrCp2Cl2 and DIBAL-H.

Recommended products

Angled septum-inlet adapter Z118206 or straight septum-inlet adapter, 6mm I.D. inlet, Z118141 or straight septum-inlet adapter, 13mm I.D. inlet, Z118192 is recommended.

septum inlet adapter

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 2 Inhalation - STOT SE 3 - Water-react 1

target_organs

Central nervous system, Nervous system

supp_hazards

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 3

flash_point_f

-9.4 °F

flash_point_c

-23 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Tomioka et al.
The Journal of organic chemistry, 76(11), 4669-4674 (2011-04-30)
Stepwise, selective DIBAL reduction of the acetonide diester derived from tartaric acid followed by the Horner-Emmons reaction effectively provided desymmetrized hydroxy mono-olefination products in a one-pot operation.
K Hirota et al.
Nucleic acids symposium series, (34)(34), 15-16 (1995-01-01)
It is reported that the diisobutylaluminum hydride (DIBALH) reduction of inosine and adenosine derivatives (1a and 1d) causes cleavage of the ribose moiety to give the corresponding 9-ribitylhypoxanthine (2a) and 9-ribityladenine (2d), respectively. The substitution effect of purine nucleosides on
Carl A Busacca et al.
The Journal of organic chemistry, 73(4), 1524-1531 (2008-01-17)
The reduction of tertiary phosphine oxides (TPOs) and sulfides with diisobutylaluminum hydride (DIBAL-H) has been studied in detail. An extensive solvent screen has revealed that hindered aliphatic ethers, such as MTBE, are optimum for this reaction at ambient temperature. Many
[The reduction of prostaglandin intermediate with diisobutylaluminium hydride].
Z Y Liu et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 17(4), 296-298 (1982-04-01)
Hideki Ishii et al.
Organic & biomolecular chemistry, 3(19), 3471-3472 (2005-09-21)
The lactone rings of ginkgolide A are converted into corresponding tetrahydrofuran moieties via DIBAL-H reduction followed by deoxygenation of the formed lactols with Et3SiH-BF3.Et2O to produce a series of lactone-free ginkgolides.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service