Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

256838

Sigma-Aldrich

Diisobutylaluminum hydride solution

1.0 M in cyclohexane

Synonym(s):

DIBAL, DIBAL-H

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHCH2]2AlH
CAS Number:
Molecular Weight:
142.22
Beilstein/REAXYS Number:
4123663
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

concentration

1.0 M in cyclohexane

density

0.781 g/mL at 25 °C

SMILES string

CC(C)C[AlH]CC(C)C

InChI

1S/2C4H9.Al.H/c2*1-4(2)3;;/h2*4H,1H2,2-3H3;;

InChI key

AZWXAPCAJCYGIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in Pd-catalyzed reductive debromination of secondary alkyl bromides. O-Debenzylation and ring opening of perbenzylated furanosides. Convenient in situ generation of HZrCp2Cl from ZrCp2Cl2 and DIBAL-H.

Recommended products

Angled septum-inlet adapter Z118206 or straight septum-inlet adapter, 6mm I.D. inlet, Z118141 or straight septum-inlet adapter, 13mm I.D. inlet, Z118192 is recommended.

replaced by

Product No.
Description
Pricing

septum inlet adapter

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hideki Ishii et al.
Organic & biomolecular chemistry, 3(19), 3471-3472 (2005-09-21)
The lactone rings of ginkgolide A are converted into corresponding tetrahydrofuran moieties via DIBAL-H reduction followed by deoxygenation of the formed lactols with Et3SiH-BF3.Et2O to produce a series of lactone-free ginkgolides.
Nobuo Tanaka et al.
Carbohydrate research, 343(15), 2675-2679 (2008-08-23)
The reductive ring-opening reaction of benzylidene-protected glucosides and mannosides such as methyl 2,3-di-O-benzyl-4,6-O-benzylidene-alpha-d-glucoside (1) and methyl 2,3-di-O-benzyl-4,6-O-benzylidene-alpha-d-mannoside (4) by using a toluene stock solution of DIBAL-H and a dichloromethane stock solution of DIBAL-H gives mainly or selectively the corresponding 2,3,4-tri-O-benzyl
V Constantinou-Kokotou et al.
The journal of peptide research : official journal of the American Peptide Society, 58(4), 325-331 (2001-10-19)
A general method for the synthesis of enantiopure non-natural alpha-amino acids is described. The key intermediate tert-butyl (2S)-2-[bis(tert-butoxycarbonyl)amino]-5-oxopentanoate was obtained from l-glutamic acid after suitable protection and selective reduction of the gamma-methyl ester group by DIBALH. Wittig reaction of this
Diisobutylaluminum hydride mediated regioselective O desilylations: access to multisubstituted cyclodextrins.
Ramprasad Ghosh et al.
Angewandte Chemie (International ed. in English), 51(7), 1548-1552 (2011-10-19)
Thomas Lecourt et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 10(12), 2960-2971 (2004-06-24)
To explain the remarkable regioselective de-O-benzylating properties of diisobutylaluminium hydride (DIBAL-H) and triisobutylaluminium (TIBAL) towards polybenzylated sugars or cyclodextrins, we propose a plausible mechanistic rationale critically involving the kinetic formation of a product-generating 2:1 Al-benzylated sugar complex. For the reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service