Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

255815

Sigma-Aldrich

Lithium tetrafluoroborate solution

1.0 M in acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiBF4
CAS Number:
Molecular Weight:
93.75
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

liquid

concentration

1.0 M in acetonitrile

density

0.852 g/mL at 25 °C

SMILES string

[Li+].F[B-](F)(F)F

InChI

1S/BF4.Li/c2-1(3,4)5;/q-1;+1

InChI key

UFXJWFBILHTTET-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Muta. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_c

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiro Ichikawa et al.
Journal of the American Chemical Society, 133(7), 2163-2169 (2011-01-29)
Thermotropic bicontinuous cubic (Cub(bi)) liquid-crystalline (LC) compounds based on a polymerizable ammonium moiety complexed with a lithium salt have been designed to obtain lithium ion-conductive all solid polymeric films having 3D interconnected ionic channels. The monomer shows a Cub(bi) phase
J S Yadav et al.
Carbohydrate research, 332(2), 221-224 (2001-07-04)
The treatment of glycals with allyltrimethylsilane in the presence of lithium tetrafluoroborate in acetonitrile gave the corresponding allyl 2,3-unsaturated C-glycosylic compounds in excellent yields with high anomeric selectivity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service