Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

255211

Sigma-Aldrich

4-(Dimethylamino)pyridinium chlorochromate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11ClCrN2O3
CAS Number:
Molecular Weight:
258.62
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

97%

form

solid

SMILES string

[Cr](=O)(=O)(Cl)[O-].[n+H]1ccc(cc1)N(C)C

InChI

1S/C7H10N2.ClH.Cr.3O/c1-9(2)7-3-5-8-6-4-7;;;;;/h3-6H,1-2H3;1H;;;;/q;;+1;;;-1

InChI key

GOOURKXKOZOZHR-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service