Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

253642

Sigma-Aldrich

4-Phosphonooxy-TEMPO hydrate, free radical

for ESR-spectroscopy

Synonym(s):

4-Phosphonooxy-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19NO5P · xH2O
CAS Number:
Molecular Weight:
252.22 (anhydrous basis)
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

mp

270 °C (dec.) (lit.)

functional group

phosphate

storage temp.

2-8°C

SMILES string

[H]O[H].CC1(C)CC(CC(C)(C)N1[O])OP(O)(O)=O

InChI

1S/C9H19NO5P.H2O/c1-8(2)5-7(15-16(12,13)14)6-9(3,4)10(8)11;/h7H,5-6H2,1-4H3,(H2,12,13,14);1H2

InChI key

CIGTZKBHXQSHHC-UHFFFAOYSA-N

General description

Spin label.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service