253359
4-Maleimido-TEMPO
for ESR-spectroscopy
Synonym(s):
4-Maleimido-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical
About This Item
Recommended Products
description
free radical
form
solid
mp
91-94 °C (lit.)
functional group
imide
maleimide
storage temp.
2-8°C
SMILES string
CC1(C)CC(CC(C)(C)N1[O])N2C(=O)C=CC2=O
InChI
1S/C13H19N2O3/c1-12(2)7-9(8-13(3,4)15(12)18)14-10(16)5-6-11(14)17/h5-6,9H,7-8H2,1-4H3
InChI key
CMNDHIFMYRPBGH-UHFFFAOYSA-N
General description
application
Biochem/physiol Actions
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service