Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

253111

Sigma-Aldrich

Borane N,N-diisopropylethylamine complex

Synonym(s):

Diisopropylethylamine borane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2NC2H5·BH3
CAS Number:
Molecular Weight:
143.08
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reagent type: reductant

refractive index

n20/D 1.46 (lit.)

mp

15-17 °C (lit.)

density

0.822 g/mL at 25 °C (lit.)

SMILES string

B.CCN(C(C)C)C(C)C

InChI

1S/C8H19N.BH3/c1-6-9(7(2)3)8(4)5;/h7-8H,6H2,1-5H3;1H3

InChI key

DWXQHSSGDGWIMP-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of modifiable bidentate systems via nitrogen-carbon rearrangement
  • Preparation of bioactive boranophosphonic acids
  • Preparation of precursors for synthesis of boronated oligonucleotide sequences
  • Synthesis of dithymidine boranophosphate via the oxathiaphospholane approach
  • Preparation of adenosine boranothiotriphosphate ribonucleoside and thymidine boranothiotriphosphate deoxyribonucleoside
  • Preparation of borane-amine complexes

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

104.0 °F - closed cup

flash_point_c

40 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service