Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

252808

Sigma-Aldrich

Octyl chloroformate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO2(CH2)7CH3
CAS Number:
Molecular Weight:
192.68
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

0.39 psi ( 20 °C)

assay

97%

form

liquid

refractive index

n20/D 1.431 (lit.)

bp

90-91 °C/11 mmHg (lit.)

density

0.984 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCCOC(Cl)=O

InChI

1S/C9H17ClO2/c1-2-3-4-5-6-7-8-12-9(10)11/h2-8H2,1H3

InChI key

VFXVAXFIFHSGNR-UHFFFAOYSA-N

Application

Octyl chloroformate has been used in preparation of:
  • octyl 4-[(tetrahydropyran-2-yl)oxy]but-2-ynoate[1]
  • octyl 4-methoxybut-2-ynoate[1]
  • octyl 4,4-diethoxybut-2-ynoate[1]
  • N-octyloxycarbonyl-R110[2]

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

168.8 °F - closed cup

flash_point_c

76 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regio-and enantioselective synthesis of allenic esters by samarium (II)-mediated reduction of propargylic compounds through dynamic kinetic protonation.
Mikami K and Yoshida A.
Tetrahedron, 57(5), 889-898 (2001)
S X Cai et al.
Bioorganic & medicinal chemistry letters, 11(1), 39-42 (2001-01-05)
N-Octyloxycarbonyl-R110 (1), with enhanced cell penetration and retention properties, was prepared from rhodamine 110. The tetrapeptide substrate N-Ac-DEVD-N'-octyloxycarbonyl-R110 (3) was prepared and shown to be efficiently cleaved by human recombinant caspase-3 and by apoptotic HL-60 cells. This substrate should prove
Tomáš Hložek et al.
Journal of analytical toxicology, 40(9), 749-753 (2016-09-04)
A simple, sensitive and robust method for simultaneous determination of antiepileptic drugs (gabapentin, pregabalin and vigabatrin) in human serum using GC-MS was developed and validated for clinical toxicology purposes. This method employs an emerging class of derivatization agents - alkyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service