Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

251798

Sigma-Aldrich

2,4-Dichlorophenyl isocyanate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NCO
CAS Number:
Molecular Weight:
188.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

form

solid

mp

60-62 °C (lit.)

functional group

chloro
isocyanate

SMILES string

Clc1ccc(N=C=O)c(Cl)c1

InChI

1S/C7H3Cl2NO/c8-5-1-2-7(10-4-11)6(9)3-5/h1-3H

InChI key

OLBJNSPBWLCTOT-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service