251496
(−)-Isopinocampheylborane TMEDA complex
Synonym(s):
(−)-N,N′-Bis(monoisopinocampheylborane)-N,N,N′,N′-tetramethylethylenediamine, (R)-Alpine-Boramine™
About This Item
Recommended Products
form
solid
mp
139-141 °C (lit.)
functional group
amine
SMILES string
CN(C)CCN(C)C.[H][C@]12C[C@H](B)[C@@H](C)[C@]([H])(C1)C2(C)C.[H][C@]34C[C@H](B)[C@@H](C)[C@]([H])(C3)C4(C)C
InChI
1S/2C10H19B.C6H16N2/c2*1-6-8-4-7(5-9(6)11)10(8,2)3;1-7(2)5-6-8(3)4/h2*6-9H,4-5,11H2,1-3H3;5-6H2,1-4H3/t2*6-,7+,8-,9-;/m00./s1
InChI key
QNDCJVKDHWRHGS-NCHGFZPDSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - Water-react 3
Storage Class
4.3 - Hazardous materials which set free flammable gases upon contact with water
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service