Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

249629

Sigma-Aldrich

Ethyl diacetoacetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO)2CHCO2C2H5
CAS Number:
Molecular Weight:
172.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.47 (lit.)

bp

104-105 °C/17 mmHg (lit.)

density

1.104 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(C(C)=O)C(C)=O

InChI

1S/C8H12O4/c1-4-12-8(11)7(5(2)9)6(3)10/h7H,4H2,1-3H3

InChI key

YMCDYRGMTRCAPZ-UHFFFAOYSA-N

General description

Ethyl diacetoacetate reacts with o-phenylenediamine to yield 2-methylbenzimidazole perchlorate[1].

Application

Ethyl diacetoacetate has been used in the preparation of 3-acetyl-6-chloro-7-hydroxy-4-methylcoumarin[2].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

185.0 °F - closed cup

flash_point_c

85 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K F Chilvers et al.
Journal of applied microbiology, 91(6), 1118-1130 (2002-02-20)
A widely used coumarin derivative is 7-hydroxy-4-methylcoumarin-beta-D-galactoside (4-methylumbelliferone-beta-D-galactoside; 4-MU-GAL). This galactoside is utilized as a substrate for the detection of the beta-galactosidase activity of coliform bacteria in water analysis. The intense fluorescence of coumarin-based molecules has enabled them to be
692. Diazepines. Part III. Some benzodiazepines.
Lloyd D, et al.
Journal of the Chemical Society, 3785-3792 (1965)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service