248843
Dimethylaluminum chloride solution
1.0 M in hexanes
Synonym(s):
Chlorodimethylaluminum, Dimethylaluminum chloride, Dimethylchloralane, Dimethylchloroalane, Dimethylchloroaluminum
About This Item
Recommended Products
form
liquid
concentration
1.0 M in hexanes
density
0.701 g/mL at 25 °C
SMILES string
C[Al](C)Cl
InChI
1S/2CH3.Al.ClH/h2*1H3;;1H/q;;+1;/p-1
InChI key
JGHYBJVUQGTEEB-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Application
recommended
signalword
Danger
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Inhalation - STOT SE 3 - Water-react 1
target_organs
Central nervous system, Nervous system
Storage Class
4.2 - Pyrophoric and self-heating hazardous materials
wgk_germany
WGK 3
flash_point_f
-9.4 °F
flash_point_c
-23 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service