Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

247316

Sigma-Aldrich

4-Chloromethyl-3,5-dimethylisoxazole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8ClNO
CAS Number:
Molecular Weight:
145.59
Beilstein/REAXYS Number:
110780
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

liquid

refractive index

n20/D 1.486 (lit.)

bp

87-88 °C/8 mmHg (lit.)

density

1.173 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1noc(C)c1CCl

InChI

1S/C6H8ClNO/c1-4-6(3-7)5(2)9-8-4/h3H2,1-2H3

InChI key

NIFAUKBQIAURIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chloromethyl-3,5-dimethylisoxazole participates in asymmetric isoxazole annulation reaction[1].

Application

4-Chloromethyl-3,5-dimethylisoxazole has been used in the preparation of:
  • 4(3-oxoalkyl)isoxazole[2]
  • (E)-3,5-dimethyl-4-(2-(thiophene-3-yl)vinyl)isoxazole[3]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

204.8 °F - closed cup

flash_point_c

96 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transformation of 4-(3-oxoalkyl) isoxazoles into pyridines.
Ohashi M, et al.
Journal of the American Chemical Society, 89(21), 5460-5461 (1967)
An asymmetric isoxazole annulation
Marron BE, et al.
Journal of Heterocyclic Chemistry, 25(4), 1067-1070 (1988)
Supramolecular assembly in side-chain conjugated thiophene copolymers.
Ananthakrishnan SJ, et al.
Royal Society of Chemistry Advances, 3(22), 8331-8340 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service