Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

247278

Sigma-Aldrich

4,4′-Oxydianiline

purified by sublimation, ≥99%

Synonym(s):

4,4′-Diaminodiphenyl ether, 4-Aminophenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O(C6H4NH2)2
CAS Number:
Molecular Weight:
200.24
Beilstein/REAXYS Number:
475735
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

10 mmHg ( 240 °C)

assay

≥99%

form

solid

purified by

sublimation

mp

188-192 °C (lit.)

SMILES string

Nc1ccc(Oc2ccc(N)cc2)cc1

InChI

1S/C12H12N2O/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12/h1-8H,13-14H2

InChI key

HLBLWEWZXPIGSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,4′-Oxydianiline was used to tailor the properties of octa(aminophenyl)-silsesquioxane cross-linked polyimide aerogels. It was also used in the preparation of novel polyimide hybrid nanocomposite containing polyhedral oligomeric silsesquioxane.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 2 - Skin Sens. 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

426.2 °F - closed cup

flash_point_c

219 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuwang Duo et al.
Journal of nanoscience and nanotechnology, 13(2), 1356-1359 (2013-05-08)
A novel polyimide (PI) hybrid nanocomposite containing polyhedral oligomeric silsesquioxane (POSS) had been prepared by copolymerization of trisilanolphenyl-POSS, 4,4'-oxydianiline (ODA), and pyromellitic dianhydride (PMDA). The AO resistance of these PI/POSS hybrid films was tested in the ground-based AO simulation facility.
Haiquan Guo et al.
ACS applied materials & interfaces, 4(10), 5422-5429 (2012-09-08)
Combinations of rigid and flexible aromatic diamines were used to tailor the properties of octa(aminophenyl)-silsesquioxane (OAPS) cross-linked polyimide aerogels. 2,2'-Dimethylbenzidine (DMBZ) or p-phenylenediamine (PPDA) was used in combination with the more-flexible diamine, 4,4'-oxydianiline (ODA). The amount of rigid diamine was
Anders Lennartson et al.
Chirality, 27(7), 425-429 (2015-06-03)
We studied the spontaneous formation of chiral crystals of four diaryl ethers, 3-phenoxybenzaldehyde, 1; 1,3-dimethyl-2-phenoxybenzene, 2; di(4-aminophenyl) ether, 3; and di(p-tolyl) ether, 4. Compounds 1, 3, and 4 form conformationally chiral molecules in the solid state, while the chirality of
4,4'-Oxydianiline.
Report on carcinogens : carcinogen profiles, 11, III210-III211 (2004-01-01)
X Chen et al.
Se pu = Chinese journal of chromatography, 17(2), 178-181 (2003-01-29)
In this paper, we introduce an HPLC qualitative method, that is, multiwavelength detection qualitative analysis method. According to Beer's law, for a certain compounds the ratio of peak areas at different wavelengths is a constant. Comparing the ratio of peak

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service