Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

243264

Supelco

Nitron

98%

Synonym(s):

1,4-Diphenyl-3-phenylamino-1,2,4-triazolium hydroxide inner salt, 3,5,6-Triphenyl-2,3,5,6-tetraazabicyclo[2.1.1]hex-1-ene, 4,5-Dihydro-2,4-diphenyl-5-(phenylimino)-1H-1,2,4-triazolium hydroxide inner salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16N4
CAS Number:
Molecular Weight:
312.37
Beilstein/REAXYS Number:
3913543
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

98%

mp

189-190 °C (dec.) (lit.)

SMILES string

[N-]1N(C=[N+](c2ccccc2)\C1=N\c3ccccc3)c4ccccc4

InChI

1S/C20H16N4/c1-4-10-17(11-5-1)21-20-22-24(19-14-8-3-9-15-19)16-23(20)18-12-6-2-7-13-18/h1-16H

Inchi Key

CWGBFIRHYJNILV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Attila Szebeni et al.
The international journal of neuropsychopharmacology, 17(10), 1579-1589 (2014-06-27)
Telomere shortening is observed in peripheral mononuclear cells from patients with major depressive disorder (MDD). Whether this finding and its biological causes impact the health of the brain in MDD is unknown. Brain cells have differing vulnerabilities to biological mechanisms
Einav Amit et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(57), 13046-13052 (2020-04-29)
N-heterocyclic carbenes (NHCs) have emerged as a unique molecular platform for the formation of self-assembled monolayers (SAMs) on various surfaces. However, active carbene formation requires deprotonation of imidazolium salt precursors, which is mostly facilitated by exposure of the salt to
Fuminori Yoneyama et al.
Bioengineered, 6(4), 209-217 (2015-04-17)
Glycerol is an interesting feedstock for biomaterials such as biofuels and bioplastics because of its abundance as a by-product during biodiesel production. Here we demonstrate glycerol metabolism in the nitrogen-fixing species Azotobacter vinelandii through metabolomics and nitrogen-free bacterial production of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service