Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

241407

Sigma-Aldrich

(S)-(−)-2-Methylbutylamine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CH2NH2
CAS Number:
Molecular Weight:
87.16
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

optical activity

[α]20/D −5.9°, neat

refractive index

n20/D 1.4126 (lit.)

bp

40-45 °C/12 mmHg (lit.)

density

0.738 g/mL at 25 °C (lit.)

SMILES string

CC[C@H](C)CN

InChI

1S/C5H13N/c1-3-5(2)4-6/h5H,3-4,6H2,1-2H3/t5-/m0/s1

InChI key

VJROPLWGFCORRM-YFKPBYRVSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

53.6 °F - closed cup

flash_point_c

12 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jay M Patel et al.
eLife, 8 (2019-05-11)
Appetite is driven by nutritional state, environmental cues, mood, and reward pathways. Environmental cues strongly influence feeding behavior, as they can dramatically induce or diminish the drive to consume food despite homeostatic state. Here, we have uncovered an excitatory neuronal
Albany Reséndiz-Mora et al.
Biochemical and biophysical research communications, 509(1), 275-280 (2018-12-26)
Non-bilayer phospholipid arrangements (NPA) are lipid associations different from the bilayer, formed by the interactions of conic anionic lipids and divalent cations that produce an inverted micelle which is inserted between the lipid layers, so the polar heads of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service