Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

237728

Sigma-Aldrich

S-Alpine-Hydride®

0.5 M in THF

Synonym(s):

Lithium B-isopinocampheyl-9-borabicyclo[3.3.1]nonyl hydride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32BLi
CAS Number:
Molecular Weight:
266.20
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

form

solid

optical activity

[α]20/D +5.5°, neat

concentration

0.5 M in THF

density

0.92 g/mL at 25 °C (lit.)

SMILES string

[Li+].C[C@@H]1[C@H](C[C@H]2C[C@@H]1C2(C)C)[BH-]3[C@@H]4CCC[C@H]3CCC4

InChI

1S/C18H32B.Li/c1-12-16-10-13(18(16,2)3)11-17(12)19-14-6-4-7-15(19)9-5-8-14;/h12-17,19H,4-11H2,1-3H3;/q-1;+1/t12-,13+,14-,15+,16-,17-,19?;/m0./s1

InChI key

JBOSJEWMOTWSER-WWPAYOJJSA-N

Legal Information

Alpine-Hydride is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B - Water-react 1

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service