Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

237620

Sigma-Aldrich

(S)-(+)-1,3-Butanediol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2CH2OH
CAS Number:
Molecular Weight:
90.12
Beilstein/REAXYS Number:
1718943
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

vapor density

3.1 (vs air)

vapor pressure

0.06 mmHg ( 20 °C)

assay

98%

optical activity

[α]20/D +30°, c = 1 in ethanol

optical purity

ee: 96% (GLC)

autoignition temp.

741 °F

refractive index

n20/D 1.440 (lit.)

bp

109 °C/14 mmHg (lit.)

density

1.005 g/mL at 25 °C (lit.)

SMILES string

C[C@H](O)CCO

InChI

1S/C4H10O2/c1-4(6)2-3-5/h4-6H,2-3H2,1H3/t4-/m0/s1

InChI key

PUPZLCDOIYMWBV-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chiral starting material.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

249.8 °F - closed cup

flash_point_c

121 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P A Pugh et al.
Animal reproduction science, 58(1-2), 9-22 (2000-03-04)
This study examined the effects of adding a macromolecule, polyvinylpyrrolidone (10% PVP) and a sugar (0.3 M trehalose) to vitrification solutions (VS) containing either one (40% ethylene glycol [EG], two (25% EG+25% DMSO) or three (20% EG+20% DMSO+10% 1, 3-butanediol
Nobuya Itoh et al.
Applied microbiology and biotechnology, 75(6), 1249-1256 (2007-04-20)
An asymmetric hydrogen-transfer biocatalyst consisting of mutated Rhodococcus phenylacetaldehyde reductase (PAR) or Leifsonia alcohol dehydrogenase (LSADH) was applied for some water-soluble ketone substrates. Among them, 4-hydroxy-2-butanone was reduced to (S)/(R)-1,3-butanediol, a useful intermediate for pharmaceuticals, with a high yield and
Biagio Velino et al.
The journal of physical chemistry. A, 115(34), 9585-9589 (2011-03-23)
The rotational spectra of five conformers of 1,3-butandiol have been measured by pulsed jet Fourier transform microwave spectroscopy. All of them are stabilized by an internal hydrogen bond and all of them have a GG' or a G'G arrangement of
Q Wu et al.
Guang pu xue yu guang pu fen xi = Guang pu, 20(5), 612-613 (2003-08-30)
In the present paper, we studied the surface structure effect of Pt electrode modified with antimony and sulfur adatoms towards 1,3-Butanediol oxidation. The results demonstrated that the modification of Sb and S both inhibited the dissociative adsorption of 1,3-Butanediol into
Allergic contact dermatitis due to 1,3-butylene glycol and glycerol.
Risa Tamagawa-Mineoka et al.
Contact dermatitis, 56(5), 297-298 (2007-04-20)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service