Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

236896

Sigma-Aldrich

Chloroform-d

99.8 atom % D

Synonym(s):

Deuterochloroform

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CDCl3
CAS Number:
Molecular Weight:
120.38
Beilstein/REAXYS Number:
1697633
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99.8 atom % D

Quality Level

assay

≥99.00%

form

liquid

packaging

pk of 10 × 1 mL

impurities

≤0.0100% water
water

refractive index

n20/D 1.444 (lit.)

bp

60.9 °C (lit.)

mp

−64 °C (lit.)

density

1.500 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]C(Cl)(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H/i1D

InChI key

HEDRZPFGACZZDS-MICDWDOJSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isotopic dilution studies of the chloroform-chloroform-d system by Raman difference spectroscopy.
Laane J and Kiefer W.
J. Chem. Phys. , 73(10), 4971-4975 (1980)
Quantitative Study of the Bonding of Chloroform-d in Various Solvents by Infrared Spectrometry1.
Lord RC, et al.
Journal of the American Chemical Society, 77(5), 1365-1368 (1955)
Infrared Intensity of the C-D Stretch of Chloroform-d in Various Solvents.
Huggins CM and Pimentel GC.
J. Chem. Phys. , 23(5), 896-898 (1955)
Yukiko Tsuji et al.
Plant biotechnology journal, 13(6), 821-832 (2015-01-13)
Bacteria-derived enzymes that can modify specific lignin substructures are potential targets to engineer plants for better biomass processability. The Gram-negative bacterium Sphingobium sp. SYK-6 possesses a Cα-dehydrogenase (LigD) enzyme that has been shown to oxidize the α-hydroxy functionalities in β-O-4-linked
Sanna L Diemer et al.
International journal of molecular sciences, 16(9), 21858-21872 (2015-09-18)
Dynamic combinatorial chemistry has emerged as a promising tool for the discovery of complex receptors in supramolecular chemistry. At the heart of dynamic combinatorial chemistry are the reversible reactions that enable the exchange of building blocks between library members in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service