Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

235105

Sigma-Aldrich

N,O-Bis(trimethylsilyl)hydroxylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiONHSi(CH3)3
CAS Number:
Molecular Weight:
177.39
Beilstein/REAXYS Number:
1920452
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.411 (lit.)

bp

78-80 °C/100 mmHg (lit.)

density

0.83 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)NO[Si](C)(C)C

InChI

1S/C6H19NOSi2/c1-9(2,3)7-8-10(4,5)6/h7H,1-6H3

InChI key

ZAEUMMRLGAMWKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,O-Bis(trimethylsilyl)hydroxylamine on reaction with 9-chloro-9-borafluorene yields 10-trimethylsilyloxy-9-aza-10-boraphenanthrene[1]. It reacts with free phenolic steroids during the analysis of conjugated estrogen tablets and injectable formulations by GLC[2].

Application

N,O-Bis(trimethylsilyl)hydroxylamine has been used as reagent for the preparation of N-(dialkyl phosphinoyl)hydroxylamines and migration of simple alkyl groups in rearrangement of their O-p-nitrobenzenesulfonates[3].

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

84.2 °F - closed cup

flash_point_c

29 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 48, 7539-7539 (1992)
Sunanda Biswas et al.
Chemical communications (Cambridge, England), 48(38), 4564-4566 (2012-04-03)
Treatment of 9-chloro-9-borafluorene with N,O-bis(trimethylsilyl)hydroxylamine results in 10-trimethylsilyloxy-9-aza-10-boraphenanthrene 6b. NMR spectroscopy shows that the expected antiaromatic 9-(trimethylsilyloxyamino)-9-borafluorene 5b rearranges to the formally aromatic phenanthrene 6b at room temperature.
G K Pillai et al.
Journal of pharmaceutical sciences, 70(9), 1072-1075 (1981-09-01)
A rapid capillary GLC method for the analysis of conjugated estrogen tablets and injectable formulations is described. The method involves the hydrolytic cleavage of the sodium sulfate ester conjugates by sulfatase enzyme. The free phenolic steroids are reacted sequentially with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service