Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

234966

Sigma-Aldrich

Methyl trimethylsilylacetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCH2CO2CH3
CAS Number:
Molecular Weight:
146.26
Beilstein/REAXYS Number:
1098961
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.414 (lit.)

bp

38-39 °C/13 mmHg (lit.)

density

0.891 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

COC(=O)C[Si](C)(C)C

InChI

1S/C6H14O2Si/c1-8-6(7)5-9(2,3)4/h5H2,1-4H3

InChI key

JIHUZDFFYVRXKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl trimethylsilylacetate on reaction with lithium diisopropylamide in THF at -78°C yields methyl lithiotrimethylsilylacetate[1]. It also undergoes Peterson olefination to yield mainly Z-isomers of steroid 17β-side chain methyl acrylates[2].

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

91.4 °F - closed cup

flash_point_c

33 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z-Isomers of Steroid 17?-Side Chain Methyl Acrylates.
Pouzar V, et al.
Collection of Czechoslovak Chemical Communications, 57(9), 1928-1936 (1992)
Conjugate addition of lithiotrimethylsilylacetate. A simple synthesis of methyl jasmonate via vicinal double alkylation.
Nishiyama H, et al.
Tetrahedron Letters, 25(23), 2487-2488 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service