232483
Diethyl 3-oxopimelate
Synonym(s):
3-Oxoheptanedioic acid diethyl ester, Diethyl 3-oxoheptanedioate
About This Item
Recommended Products
form
liquid
refractive index
n20/D 1.447 (lit.)
bp
130-132 °C/0.5 mmHg (lit.)
density
1.084 g/mL at 25 °C (lit.)
functional group
ester
ketone
SMILES string
CCOC(=O)CCCC(=O)CC(=O)OCC
InChI
1S/C11H18O5/c1-3-15-10(13)7-5-6-9(12)8-11(14)16-4-2/h3-8H2,1-2H3
InChI key
RTMQRCLOAACETL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Regioselective one-pot synthesis of 1,3,4-trisubstituted-1H-pyrazoles
- Synthesis of lupinine via stereoselective multicomponent condensation
- Solvent-free condensation of β-keto esters with amidoximes
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service