Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23195

Sigma-Aldrich

4-tert-Butylcatechol solution

≥98% (calc. to the dried substance, GC), liquid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O2
CAS Number:
Molecular Weight:
166.22
Beilstein/REAXYS Number:
2043335
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥98% (calc. to the dried substance, GC)

form

liquid

impurities

≤15% methanol

loss

≤15% loss on drying, 24 h (vacuum)

SMILES string

CC(C)(C)c1ccc(O)c(O)c1

InChI

1S/C10H14O2/c1-10(2,3)7-4-5-8(11)9(12)6-7/h4-6,11-12H,1-3H3

InChI key

XESZUVZBAMCAEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 1

target_organs

Eyes,Central nervous system

wgk_germany

WGK 3

ppe

Faceshields, Gloves, Goggles


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Contact sensitivity to para-tertiary-butylcatechol in an artificial limb.
A W Macfarlane et al.
Contact dermatitis, 22(1), 56-57 (1990-01-01)
Martín A Fernández-Baldo et al.
Talanta, 79(3), 681-686 (2009-07-07)
Botrytis cinerea is a plant-pathogenic fungus that produces the disease known as grey mould in a wide variety of agriculturally important hosts in many countries. This paper describes the development of an immunosensor coupled to carbon-based screen-printed electrodes (SPCE) modified
T Kawashima et al.
The Journal of investigative dermatology, 82(1), 53-56 (1984-01-01)
4-Tertiary butyl catechol (TBC) causes depigmentation in humans and animals and stimulates formation of pheomelanosomes. In this study, we investigated the effects of noncytotoxic doses of TBC on glutathione S-transferase (GST) activity in the skin of Uscd strain mice and
María José Peñalver et al.
Biochimica et biophysica acta, 1597(1), 140-148 (2002-05-16)
The stoichiometry of oxygen consumption during tyrosinase-catalyzed oxidation of an o-diphenol (4-tert-butylcatechol, TBC) and a monophenol (4-tert-butylphenol, TBP) has been determined. At high [substrate]/[enzyme] ratios, in the case of o-diphenols, the stoichiometry of the enzyme-catalyzed reaction was always 1 O(2)/2
Sirley V Pereira et al.
The Analyst, 136(22), 4745-4751 (2011-10-11)
In this article we report the development of an integrated microfluidic system coupled to a screen-printed carbon electrode (SPCE) applied to the quantitative determination of IgG specific antibodies present in serum samples of patients that suffer from Chagas disease. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service