Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23021

Sigma-Aldrich

2-(2-Chloroethoxy)tetrahydro-2H-pyran

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13ClO2
CAS Number:
Molecular Weight:
164.63
Beilstein/REAXYS Number:
1280441
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥97.0%

refractive index

n20/D 1.459

bp

80 °C/8 mmHg (lit.)

density

1.115 g/mL at 20 °C (lit.)

SMILES string

ClCCOC1CCCCO1

InChI

1S/C7H13ClO2/c8-4-6-10-7-3-1-2-5-9-7/h7H,1-6H2

InChI key

ZVVQFPGYDBUGQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(2-Chloroethoxy)tetrahydro-2H-pyran is a building block for introducing a protected 2-hydroxyethyl group[1][2][3].

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

149.0 °F - closed cup

flash_point_c

65 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H.G. Lohr et al.
Chemische Berichte, 118, 914-914 (1985)
Michael Klein et al.
PloS one, 6(6), e20789-e20789 (2011-06-24)
Highly selective, cell-permeable and fast-acting inhibitors of individual kinases are sought-after as tools for studying the cellular function of kinases in real time. A combination of small molecule synthesis and protein mutagenesis, identified a highly potent inhibitor (1-Isopropyl-3-(phenylethynyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine) of a
B. Son et al.
Synthesis, 776-776 (1984)
H Ikuta et al.
Journal of medicinal chemistry, 30(11), 1995-1998 (1987-11-01)
A series of 3-(3,5-di-tert-butyl-4-hydroxybenzylidene)pyrrolidin-2-ones was synthesized and evaluated as candidate antiinflammatory/analgesic agents as well as dual inhibitors of prostaglandin and leukotriene synthesis. Some compounds that showed dual inhibitory activity were found to possess equipotent antiinflammatory activities to indomethacin, with reduced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service