Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

229245

Sigma-Aldrich

Tributyltin methoxide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3SnOCH3
CAS Number:
Molecular Weight:
321.09
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

assay

97%

form

liquid

reaction suitability

core: tin

refractive index

n20/D 1.472 (lit.)

bp

97-97.5 °C/0.06 mmHg (lit.)

density

1.115 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(CCCC)OC

InChI

1S/3C4H9.CH3O.Sn/c3*1-3-4-2;1-2;/h3*1,3-4H2,2H3;1H3;/q;;;-1;+1

InChI key

KJGLZJQPMKQFIK-UHFFFAOYSA-N

application

For a review on tin reagents in organic synthesis see Aldrichimica Acta.[1]

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

210.2 °F - closed cup

flash_point_c

99 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 17, 75-75 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service