Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

229024

Sigma-Aldrich

R-Alpine-Hydride®

0.5 M in THF

Synonym(s):

Lithium B-isopinocampheyl-9-borabicyclo[3.3.1]nonyl hydride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32BLi
CAS Number:
Molecular Weight:
266.20
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

form

liquid

optical activity

[α]20/D −8.5°

concentration

0.5 M in THF

density

0.919 g/mL at 25 °C (lit.)

SMILES string

[Li+].C[C@H]1[C@@H](C[C@@H]2C[C@H]1C2(C)C)[BH-]3[C@@H]4CCC[C@H]3CCC4

InChI

1S/C18H32B.Li/c1-12-16-10-13(18(16,2)3)11-17(12)19-14-6-4-7-15(19)9-5-8-14;/h12-17,19H,4-11H2,1-3H3;/q-1;+1/t12-,13+,14-,15+,16-,17-,19?;/m1./s1

InChI key

JBOSJEWMOTWSER-ZEBZFEJDSA-N

Other Notes

Made from (+)-α-pinene

Legal Information

Alpine-Hydride is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

target_organs

Respiratory system

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

1.4 °F

flash_point_c

-17 °C

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service