Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

228508

Sigma-Aldrich

Lithium

shot, 99%, 4-16 mesh, in mineral oil

Synonym(s):

Lithium atom, Lithium element

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
Li
CAS Number:
Molecular Weight:
6.94
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

99%

form

shot

reaction suitability

reagent type: reductant

resistivity

9.446 μΩ-cm, 20°C

weight

Unit weight is weight of contained metal

particle size

4-16 mesh

bp

1342 °C (lit.)

mp

180 °C (lit.)

density

0.534 g/mL at 25 °C (lit.)

SMILES string

[Li]

InChI

1S/Li

InChI key

WHXSMMKQMYFTQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged under argon

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B - Water-react 1

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enikö Zörgö et al.
PLoS genetics, 9(3), e1003388-e1003388 (2013-04-05)
The number of chromosome sets contained within the nucleus of eukaryotic organisms is a fundamental yet evolutionarily poorly characterized genetic variable of life. Here, we mapped the impact of ploidy on the mitotic fitness of baker's yeast and its never
U Eismann et al.
Optics express, 21(7), 9091-9102 (2013-04-11)
We present an all-solid-state laser source emitting up to 2.1 W of single-frequency light at 671 nm developed for laser cooling of lithium atoms. It is based on a diode-pumped, neodymium-doped orthovanadate (Nd:YVO(4)) ring laser operating at 1342 nm. Optimization
Dongyun Chen et al.
Nanoscale, 5(17), 7890-7896 (2013-07-16)
Two-dimensional nanosheets can leverage on their open architecture to support facile insertion and removal of Li(+) as lithium-ion battery electrode materials. In this study, two two-dimensional nanosheets with complementary functions, namely nitrogen-doped graphene and few-layer WS2, were integrated via a
Jian Jiang et al.
Nanoscale, 5(17), 8105-8113 (2013-07-26)
Controlled integration of multiple semiconducting oxides into each single unit of ordered nanotube arrays is highly desired in scientific research for the realization of more attractive applications. We herein report a diffusion-controlled solid-solid route to evolve simplex Co(CO3)0.5(OH)0.11H2O@TiO2 core-shell nanowire
Chien-Hsiun Chen et al.
The New England journal of medicine, 370(2), 119-128 (2013-12-27)
Lithium has been a first-line choice for maintenance treatment of bipolar disorders to prevent relapse of mania and depression, but many patients do not have a response to lithium treatment. We selected subgroups from a sample of 1761 patients of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service