22710
Quinoxaline
≥95.0%
Synonym(s):
1,4-Benzodiazine, Benzo[a]pyrazine, Benzopyrazine
About This Item
Recommended Products
assay
≥95.0%
form
solid
bp
220-223 °C (lit.)
mp
29-32 °C (lit.)
solubility
alcohol: freely soluble(lit.)
benzene: freely soluble(lit.)
diethyl ether: freely soluble(lit.)
water: freely soluble(lit.)
density
1.124 g/mL at 25 °C (lit.)
SMILES string
c1ccc2nccnc2c1
InChI
1S/C8H6N2/c1-2-4-8-7(3-1)9-5-6-10-8/h1-6H
InChI key
XSCHRSMBECNVNS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service