223875
α-Hydroxyhippuric acid
98%
Synonym(s):
(Benzoylamino)hydroxyacetic acid
About This Item
Recommended Products
assay
98%
mp
211 °C (dec.) (lit.)
functional group
amide
carboxylic acid
hydroxyl
phenyl
SMILES string
OC(NC(=O)c1ccccc1)C(O)=O
InChI
1S/C9H9NO4/c11-7(10-8(12)9(13)14)6-4-2-1-3-5-6/h1-5,8,12H,(H,10,11)(H,13,14)
InChI key
GCWCVCCEIQXUQU-UHFFFAOYSA-N
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service