Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

220604

Sigma-Aldrich

2-Methyl-9-acridinecarboxaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11NO
CAS Number:
Molecular Weight:
221.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

145-146 °C (lit.)

SMILES string

[H]C(=O)c1c2ccccc2nc3ccc(C)cc13

InChI

1S/C15H11NO/c1-10-6-7-15-12(8-10)13(9-17)11-4-2-3-5-14(11)16-15/h2-9H,1H3

Inchi Key

PIBZGMVFXCKDER-UHFFFAOYSA-N

Application

2-Methyl-9-acridinecarboxaldehyde was used:
  • in cleavage assay of rRNA by denaturing gel electrophoresis
  • in the synthesis of peptide-acridine/acridone conjugates, potential anticancer, antiviral and antiprion drugs

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C H Tung et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(15), 7114-7118 (1992-08-01)
A series of peptide-acridine conjugates was designed and synthesized, based on three features of the proposed catalytic mechanism of RNase A: 2'-proton abstraction by His-12, proton donation to the leaving 5'-oxygen by His-119, and stabilization of the pentacoordinated phosphorous transition
M Kukowska-Kaszuba et al.
Current medicinal chemistry, 14(29), 3079-3104 (2008-01-29)
This paper consists in information about structure, synthesis and biological activity of peptide-acridine/acridone conjugates which are potential anticancer, antiviral and antiprion drugs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service