Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

219282

Sigma-Aldrich

(+)-Emetine dihydrochloride hydrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H40N2O4 · 2HCl · xH2O
CAS Number:
Molecular Weight:
553.56 (anhydrous basis)
EC Number:
UNSPSC Code:
12352203
PubChem Substance ID:

assay

97%

optical activity

[α]25/D +9.0°, c = 1 in H2O

mp

235 °C (dec.) (lit.)

SMILES string

O.Cl.Cl.CC[C@@H]1CN2CCc3cc(OC)c(OC)cc3[C@H]2C[C@H]1C[C@@H]4NCCc5cc(OC)c(OC)cc45

InChI

1S/C29H40N2O4.2ClH.H2O/c1-6-18-17-31-10-8-20-14-27(33-3)29(35-5)16-23(20)25(31)12-21(18)11-24-22-15-28(34-4)26(32-2)13-19(22)7-9-30-24;;;/h13-16,18,21,24-25,30H,6-12,17H2,1-5H3;2*1H;1H2/t18-,21-,24+,25-;;;/m1.../s1

Inchi Key

IZTPMTAWOCEKKM-MZRUSFJHSA-N

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service