Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

219088

Sigma-Aldrich

Berberine hydrochloride hydrate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18ClNO4 · HCl · xH2O
CAS Number:
Molecular Weight:
408.28 (anhydrous basis)
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

assay

99%

InChI

1S/C20H18NO4.ClH.H2O/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2;;/h3-4,7-10H,5-6,11H2,1-2H3;1H;1H2

InChI key

RSHKTKUTIXIHFF-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cemile Selin Aksoy et al.
Microbial pathogenesis, 149, 104542-104542 (2020-10-04)
A plethora of natural products emerges as attractive molecules in the struggle against antibiotic resistance. These molecules impose their bioactivities not only alone but also in combinations as well, which further enhances their effects. Berberine is a well-known isoquinoline alkaloid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service