Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

213853

Sigma-Aldrich

α-Acetamidocinnamic acid

98%

Synonym(s):

N-Acetyldehydrophenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=C(NHCOCH3)COOH
CAS Number:
Molecular Weight:
205.21
Beilstein/REAXYS Number:
1912549
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

mp

188-190 °C (lit.)

solubility

methanol: soluble 100 mg/mL, clear, yellow-green
95% ethanol: soluble 5%, clear, yellow

SMILES string

CC(=O)N\C(=C/c1ccccc1)C(O)=O

InChI

1S/C11H11NO3/c1-8(13)12-10(11(14)15)7-9-5-3-2-4-6-9/h2-7H,1H3,(H,12,13)(H,14,15)/b10-7-

InChI key

XODAOBAZOQSFDS-YFHOEESVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Asymmeteric hydrogenation of α-acetamidocinnamic acid with chiral rhodium (I) complexes on charcoal has been reported[1]. α-Acetamidocinnamic acid undergoes asymmetric hydrogenation in ionic liquid catalyzed by rhodium complex to yield (S)-phenylalanine[2].

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Choong Eui Song
Chemical communications (Cambridge, England), 9(9), 1033-1043 (2004-04-30)
Recent developments in the enantioselective chemo- and bio-catalysis in ionic liquids are reviewed. In many cases, the use of ionic liquids provides many advantages over reactions in conventional organic solvents in terms of activity, enantioselectivity, stability and the reusability of
Asymmetric hydrogenation of a-acetamidocinnamic acid with chiral rhodium complexes of DIOP and BPPM on charcoal.
Masami I, et al.
Chemical & Pharmaceutical Bulletin, 31(10), 3371-3376 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service