Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

211788

Sigma-Aldrich

4-Nitrobenzotrifluoride

98%

Synonym(s):

4-Nitro-α,α,α-trifluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CF3
CAS Number:
Molecular Weight:
191.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

form

solid

bp

81-83 °C/10 mmHg (lit.)

mp

38-40 °C (lit.)

functional group

fluoro
nitro

SMILES string

[O-][N+](=O)c1ccc(cc1)C(F)(F)F

InChI

1S/C7H4F3NO2/c8-7(9,10)5-1-3-6(4-2-5)11(12)13/h1-4H

InChI key

XKYLCLMYQDFGKO-UHFFFAOYSA-N

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

190.4 °F - closed cup

flash_point_c

88 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Perfluoroalkylene-activated poly (aryl ether) synthesis.
Labadie JW and Hedrick JL.
Macromolecules, 23(26), 5371-5373 (1990)
Aromatic Compounds with Fluorinated Side Chains.
Fialkov YA and Yagupolski LM.
Syntheses of Fluoroorganic Compounds, 233-289 (1985)
Lijuan Lan et al.
Environmental technology, 40(12), 1615-1623 (2018-01-11)
Carbon nanotubes (CNTs), activated carbon (AC), graphene, and aluminum oxide (Al2O3) supported 5% Pd catalysts were prepared by the conventional impregnation method, and catalytic activity was tested in the hydrogenation of 4-chlorophenol (4-CP) and nitrobenzene (NB) under ambient conditions (313 K

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service