Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

209600

Sigma-Aldrich

Methyl cyclohexylacetate

99%

Synonym(s):

Methyl cyclohexaneacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH2CO2CH3
CAS Number:
Molecular Weight:
156.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

form

liquid

refractive index

n20/D 1.446 (lit.)

bp

201 °C (lit.)

solubility

water: soluble 0.3 g/L at 30 °C

density

0.951 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

COC(=O)CC1CCCCC1

InChI

1S/C9H16O2/c1-11-9(10)7-8-5-3-2-4-6-8/h8H,2-7H2,1H3

InChI key

IMXBRVLCKXGWSS-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

177.8 °F

flash_point_c

81 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of perfluorobicyclic ethers [1]. The electrochemical fluorination of cycloalkyl-substituted carboxylic acids.
Abe T, et al.
Journal of Fluorine Chemistry, 23(2), 123-146 (1983)
Acyl hydrazines as precursors to acyl radicals.
Braslau R, et al.
Tetrahedron, 58(27), 5513-5523 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service