Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

205834

Sigma-Aldrich

Palladium black

Synonym(s):

Precipitated palladium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pd
CAS Number:
Molecular Weight:
106.42
EC Number:
MDL number:
UNSPSC Code:
12141733
PubChem Substance ID:

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

SMILES string

[Pd]

InChI

1S/Pd

Inchi Key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Palladium black can be used as a catalyst with a high surface area of ~47 m2g−1 in a variety of sustainable applications.[1] It can be used as an anode catalyst that shows good performance and can be used in polymer exchange membrane (PEM) based fuel cells.[2] It can also be used as an alternative catalyst to carbon-supported palladium for the hydrogenolysis reactions.[3]

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Freestanding palladium nanosheets with plasmonic and catalytic properties
Huang X, et al.
Nature Nanotechnology, 6(1), 28-28 (2011)
Palladium black as model catalyst in the hydrogenolysis of CCl2F2 (CFC-12) into CH2F2 (HFC-32)
Van de Sandt E, et al.
Applied Catalysis A: General, 155(1), 59-73 (1997)
The behavior of palladium catalysts in direct formic acid fuel cells
Zhu Y, et al.
Journal of Power Sources, 139(1-2), 15-20 (2005)
Ling Li et al.
Nature chemistry, 5(7), 607-612 (2013-06-22)
The development of transition metal-catalysed cross-coupling reactions has greatly influenced the manner in which the synthesis of complex organic molecules is approached. A wide variety of methods are now available for the formation of C(sp(2))-C(sp(2)) bonds, and more recent work
V Volarevic et al.
Journal of B.U.ON. : official journal of the Balkan Union of Oncology, 18(1), 131-137 (2013-04-25)
As novel therapeutic agents relevant to colon cancer therapy are explored continuously, we tested 4 R2edda-type ligand precursors O,O'-dialkyl esters of (S,S)-ethylenediamine-N,N'-di-2-(4-methyl)pentanoic acid (L1.2HCl-L4.2HCl) and corresponding palladium(II) and platinum(II) complexes against the human colon cancer cell lines CaCo-2, SW480 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service