Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

202835

Sigma-Aldrich

Bismuth

pieces, elongated, 99.999%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Bi
CAS Number:
Molecular Weight:
208.98
EC Number:
MDL number:
UNSPSC Code:
12141706
PubChem Substance ID:

vapor pressure

<0.1 mmHg ( 20 °C)

assay

99.999%

form

pieces, elongated

resistivity

129 μΩ-cm, 20°C

bp

1560 °C (lit.)

mp

271 °C (lit.)

density

9.8 g/mL at 25 °C (lit.)

SMILES string

[Bi]

InChI

1S/Bi

InChI key

JCXGWMGPZLAOME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philip C Andrews et al.
Chemical communications (Cambridge, England), 49(28), 2870-2872 (2013-03-02)
Reaction of Bi(O(t)Bu)3 with aspirin (acetylsalicylic acid = aspH) in dry toluene results in the bismuth(III) complex, [Bi(O2C(C6H4)OAc)3]∞ 1 (O2C(C6H4)OAc = asp), minimum inhibitory concentration (MIC) against Helicobacter pylori ≥ 6.25 μg mL(-1), while the inclusion of a stoichiometric equivalent
Julie Pannequin et al.
The Journal of biological chemistry, 279(4), 2453-2460 (2003-10-08)
Although bismuth salts have been used for over two centuries for the treatment of various gastrointestinal disorders, the mechanism of their therapeutic action remains controversial. Because gastrins bind two trivalent ferric ions with high affinity, and because ferric ions are
Jia-Tao Sun et al.
Physical review letters, 109(24), 246804-246804 (2013-02-02)
Scanning tunnelling microscopy and spectroscopy experiments complemented by first-principles calculations have been conducted to study the electronic structure of 4 monolayer Bi(110) nanoribbons on epitaxial graphene on silicon carbide [4H-SiC(0001)]. In contrast with the semimetal property of elemental bismuth, an
A Slikkerveer et al.
Medical toxicology and adverse drug experience, 4(5), 303-323 (1989-09-01)
Inorganic bismuth salts are poorly soluble in water: solubility is influenced by the acidity of the medium and the presence of certain compounds with (hydr)oxy or sulfhydryl groups. The analysis of bismuth in biological material is not standardised and is
Shunbin Lu et al.
Optics express, 21(2), 2072-2082 (2013-02-08)
The third order nonlinear optical property of Bi₂Se₃, a kind of topological insulator (TI), has been investigated under femto-second laser excitation. The open and closed aperture Z-scan measurements were used to unambiguously distinguish the real and imaginary part of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service