Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

202568

Sigma-Aldrich

Ammonium aluminum sulfate dodecahydrate

99.995% trace metals basis

Synonym(s):

Aluminum ammonium sulfate dodecahydrate, Ammonium alum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlNH4(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
453.33
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

assay

99.995% trace metals basis

form

crystals and lumps

mp

93.5 °C (lit.)

SMILES string

[Al+3].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H][N+]([H])([H])[H].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Al.H3N.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;1H3;2*(H2,1,2,3,4);12*1H2/q+3;;;;;;;;;;;;;;;/p-3

Inchi Key

WZUKKIPWIPZMAS-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

[A method for determining ammonium alum-based aerosol-forming dusts in mine air].
R V Gorskaia et al.
Gigiena truda i professional'nye zabolevaniia, (2)(2), 46-47 (1989-01-01)
David A Stevens et al.
Chemical senses, 33(5), 405-413 (2008-03-05)
Three multidimensional scaling studies were conducted to compare the taste qualities evoked from electrical and chemical stimulation, including ferrous sulfate as a typical "metallic" taste stimulus. Bipolar, anodal, and cathodal stimulation were delivered by 1.5- or 3-V batteries applied to
Yehua Shen et al.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine, 25(3), 541-551 (2012-03-08)
Matrix metalloproteinases (MMPs) are zinc-dependent endopeptidases and key regulators for many physiological and pathological functions. The MMP inhibitors have been shown to modulate diseases such as cancer, inflammation, and cardiovascular diseases. In this paper we tracked the MMP inhibitory activities
V Hostomská et al.
Journal of hazardous materials, 147(1-2), 342-349 (2007-02-20)
Experiments with cooling crystallization of ammonium alum, (NH(4)Al(SO(4))2.12H(2)O), were performed with concentrated multicomponent acidic solutions (originating from underground uranium leaching in Stráz pod Ralskem area, Czech Republic, and containing as the principal components Al3+, NH4+, and SO4(2-) ions) as well
Mutsuko Hirata-Koizumi et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 49(9), 1948-1959 (2011-05-17)
Aluminium ammonium sulfate (AAS) was tested for reproductive/developmental toxicity in a two-generation study. Male and female rats were continuously given AAS in drinking water at 0, 50, 500 or 5000 ppm. Water consumption was decreased in all AAS-treated groups, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service