200662
Ethyl cellulose
viscosity 100 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 49% ethoxyl
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
powder
autoignition temp.
698 °F
concentration
≥49.0%
extent of labeling
49% ethoxyl
refractive index
n20/D 1.47 (lit.)
viscosity
100 cP, 5 % in toluene/ethanol 80:20(lit.)
density
1.14 g/mL at 25 °C (lit.)
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ghulam Murtaza
Acta poloniae pharmaceutica, 69(1), 11-22 (2012-05-12)
Ethylcellulose (EC) based microencapsulated drug delivery systems are being extensively studied throughout the world for achieving extended drug release and protecting the core substance from degradation. The in vitro evaluation of EC microcapsules have elucidated that their particle characteristics are
H V Adikane et al.
Applied biochemistry and biotechnology, 169(3), 1026-1038 (2013-01-09)
The chemical modification of developed ethyl cellulose-based membrane was carried out to make it suitable for bioseparation. The different reagents were used for the modification of membrane to couple protein A (PA) to study the purification of immunoglobulin G (IgG)
Hanzhou Feng et al.
Colloids and surfaces. B, Biointerfaces, 103, 530-537 (2012-12-26)
Genipin-crosslinked complex microspheres made of the combination of two polymers, ethyl cellulose and chitosan, were prepared by spray drying method. Rifabutin, an anti-tuberculosis agent was used as a model drug. Effects of various specifications of ethyl cellulose and chitosan, different
Christianne Mounir Zaki Rizkalla et al.
Pharmaceutical development and technology, 18(1), 196-209 (2012-06-06)
The systemic effect of hydroxyzine hydrochloride following its oral administration or topical application is associated with non compliant anticholinergic effect. Subsequently, the present study aims to prepare microcapsules loaded with hydroxyzine hydrochloride enabling its controlled release into the skin and
N Suwannateep et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 82(3), 485-490 (2012-09-08)
The phytochemical curcumin possesses antioxidant activity; however, it becomes unstable after being exposed to light or heat or loses activity during storage. This is especially important when curcumin is applied to the skin within a cosmetic or pharmaceutical formulation, since
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service