Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

200581

Sigma-Aldrich

Poly(ethylene-co-ethyl acrylate)

ethyl acrylate 18 wt. %, beads, melt index 20 g/10 min

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH2)x[CH2CH(CO2C2H5)]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

beads

melt index

20 g/10 min

composition

ethyl acrylate, 18 wt. %

inherent viscosity

0.78 dL/g(lit.)

transition temp

softening point 116 °C

density

0.93 g/mL at 25 °C (lit.)

SMILES string

C=C.CCOC(=O)C=C

InChI

1S/C5H8O2.C2H4/c1-3-5(6)7-4-2;1-2/h3H,1,4H2,2H3;1-2H2

InChI key

CGPRUXZTHGTMKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Horowitz et al.
Vox sanguinis, 48(3), 150-155 (1985-01-01)
The red blood cells of blood stored in containers made of polyvinylchloride (PVC) film are osmotically more stable and lose on average about 1/3 less hemoglobin than when blood is stored in another plastic [poly-(ethylene-co-ethyl acrylate); EEA]. The stability of
B Horowitz et al.
Vox sanguinis, 48(3), 143-149 (1985-01-01)
The biochemical status of whole blood stored in containers fabricated of ethylene ethylacrylate (EEA) film was monitored at several times during 4 weeks of storage at 4 degrees C. Fifteen biochemical indicators were studied to reflect on erythrocyte integrity, cellular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service