Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

199486

Sigma-Aldrich

2-Bromo-2′,4′-dimethoxyacetophenone

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3COCH2Br
CAS Number:
Molecular Weight:
259.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

form

powder

mp

102-104 °C (lit.)

SMILES string

COc1ccc(C(=O)CBr)c(OC)c1

InChI

1S/C10H11BrO3/c1-13-7-3-4-8(9(12)6-11)10(5-7)14-2/h3-5H,6H2,1-2H3

InChI key

PKVBZABQCCQHLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Bromo-2′,4′-dimethoxyacetophenone inhibited the activity of papain[1].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P I Clark et al.
European journal of biochemistry, 84(1), 293-299 (1978-03-01)
Photolysis of papain which had been inhibited with 2-bromo-2',4'-dimethoxyacetophenone regenerated papain, but also formed [deltaSer25]-papain (i.e. papain in which the active-site cysteine residue 25 was replaced by dehydroserine) via the intermediate dehydrocysteine analogue, [deltaCys25]-papain. Reduction with sodium borohydride gave [Ser25]papain.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service